missing translation for 'onlineSavingsMsg'
Learn More

2,2'-Bipyridine-6,6'-dicarboxylic Acid 98.0+%, TCI America™

Catalog No. p-7123007
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B35331G 1 g
B35335G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B35331G Supplier TCI America Supplier No. B35331G

Chemical Identifiers

CAS 4479-74-7
Molecular Formula C12H8N2O4
Molecular Weight (g/mol) 244.21
MDL Number MFCD11042476
InChI Key DASAXWLMIWDYLQ-UHFFFAOYSA-N
PubChem CID 9959961
IUPAC Name [2,2'-bipyridine]-6,6'-dicarboxylic acid
SMILES OC(=O)C1=CC=CC(=N1)C1=NC(=CC=C1)C(O)=O

Specifications

Melting Point 288°C
Color White-Yellow
Quantity 1 g
Formula Weight 244.21
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,2′-Bipyridine-6,6′-dicarboxylic Acid