missing translation for 'onlineSavingsMsg'
Learn More

2,2'-Bipyridine-5,5'-dicarboxylic Acid 98.0+%, TCI America™

Catalog No. B35021G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B35021G 1 g
B35025G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B35021G Supplier TCI America Supplier No. B35021G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1802-30-8 | C12H8N2O4 | 244.206 g/mol

Chemical Identifiers

CAS 1802-30-8
Molecular Formula C12H8N2O4
Molecular Weight (g/mol) 244.206
MDL Number MFCD01318320
InChI Key KVQMUHHSWICEIH-UHFFFAOYSA-N
PubChem CID 192744
IUPAC Name 6-(5-carboxypyridin-2-yl)pyridine-3-carboxylic acid
SMILES C1=CC(=NC=C1C(=O)O)C2=NC=C(C=C2)C(=O)O

Specifications

Melting Point 365°C
Color White-Yellow
Quantity 1 g
Formula Weight 244.21
Percent Purity ≥98.0% (GC,T)
Grade Laboratory
Physical Form Crystalline Powder
Chemical Name or Material 2,2′-Bipyridine-5,5′-dicarboxylic Acid