missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,2'-Bipyridine-4,4'-dicarboxylic Acid 96.0+%, TCI America™
$29.83 - $102.52
Chemical Identifiers
| CAS | 6813-38-3 |
|---|---|
| Molecular Formula | C12H6N2O4 |
| Molecular Weight (g/mol) | 242.19 |
| MDL Number | MFCD00015430 |
| InChI Key | FXPLCAKVOYHAJA-UHFFFAOYSA-L |
| Synonym | 2,2'-bipyridine-4,4'-dicarboxylic acid, 2,2'-bipyridyl-4,4'-dicarboxylic acid, 4,4'-dicarboxy-2,2'-bipyridine, 2,2'-biisonicotinic acid, 2,2'-dipyridyl-4,4'-dicarboxylic acid, 2,2'-bipyridine-4,4'-dicarboxylicacid, 2-4-carboxypyridin-2-yl pyridine-4-carboxylic acid, 4,4-dicarboxy-2,2-bipyridine, 2-4-carboxy-2-pyridyl pyridine-4-carboxylic acid |
| PubChem CID | 688094 |
| IUPAC Name | [2,2'-bipyridine]-4,4'-dicarboxylate |
| SMILES | [O-]C(=O)C1=CC=NC(=C1)C1=CC(=CC=N1)C([O-])=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
B1876100MG
|
TCI America
B1876100MG |
100 mg |
Each for $29.83
|
|
|||||
|
B18761G
|
TCI America
B18761G |
1 g |
Each for $102.52
|
|
|||||
Chemical Identifiers
| 6813-38-3 | |
| 242.19 | |
| FXPLCAKVOYHAJA-UHFFFAOYSA-L | |
| 688094 | |
| [O-]C(=O)C1=CC=NC(=C1)C1=CC(=CC=N1)C([O-])=O |
| C12H6N2O4 | |
| MFCD00015430 | |
| 2,2'-bipyridine-4,4'-dicarboxylic acid, 2,2'-bipyridyl-4,4'-dicarboxylic acid, 4,4'-dicarboxy-2,2'-bipyridine, 2,2'-biisonicotinic acid, 2,2'-dipyridyl-4,4'-dicarboxylic acid, 2,2'-bipyridine-4,4'-dicarboxylicacid, 2-4-carboxypyridin-2-yl pyridine-4-carboxylic acid, 4,4-dicarboxy-2,2-bipyridine, 2-4-carboxy-2-pyridyl pyridine-4-carboxylic acid | |
| [2,2'-bipyridine]-4,4'-dicarboxylate |
Specifications
| 6813-38-3 | |
| C12H6N2O4 | |
| 100 mg | |
| FXPLCAKVOYHAJA-UHFFFAOYSA-L | |
| [2,2'-bipyridine]-4,4'-dicarboxylate | |
| 688094 | |
| ≥96.0% (HPLC,T) | |
| 2,2′-Bipyridine-4,4′-dicarboxylic Acid |
| White-Yellow | |
| MFCD00015430 | |
| 2,2'-bipyridine-4,4'-dicarboxylic acid, 2,2'-bipyridyl-4,4'-dicarboxylic acid, 4,4'-dicarboxy-2,2'-bipyridine, 2,2'-biisonicotinic acid, 2,2'-dipyridyl-4,4'-dicarboxylic acid, 2,2'-bipyridine-4,4'-dicarboxylicacid, 2-4-carboxypyridin-2-yl pyridine-4-carboxylic acid, 4,4-dicarboxy-2,2-bipyridine, 2-4-carboxy-2-pyridyl pyridine-4-carboxylic acid | |
| [O-]C(=O)C1=CC=NC(=C1)C1=CC(=CC=N1)C([O-])=O | |
| 242.19 | |
| 244.21 | |
| Crystalline Powder |
Safety and Handling
TSCA : No