missing translation for 'onlineSavingsMsg'
Learn More

2,2'-Bipyridine-4,4'-dicarboxylic Acid 96.0+%, TCI America™

Catalog No. B18761G
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B18761G 1 g
B1876100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B18761G Supplier TCI America Supplier No. B18761G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 6813-38-3 | C12H6N2O4 | 242.19 g/mol

Chemical Identifiers

CAS 6813-38-3
Molecular Formula C12H6N2O4
Molecular Weight (g/mol) 242.19
MDL Number MFCD00015430
InChI Key FXPLCAKVOYHAJA-UHFFFAOYSA-L
Synonym 2,2'-bipyridine-4,4'-dicarboxylic acid, 2,2'-bipyridyl-4,4'-dicarboxylic acid, 4,4'-dicarboxy-2,2'-bipyridine, 2,2'-biisonicotinic acid, 2,2'-dipyridyl-4,4'-dicarboxylic acid, 2,2'-bipyridine-4,4'-dicarboxylicacid, 2-4-carboxypyridin-2-yl pyridine-4-carboxylic acid, 4,4-dicarboxy-2,2-bipyridine, 2-4-carboxy-2-pyridyl pyridine-4-carboxylic acid
PubChem CID 688094
IUPAC Name [2,2'-bipyridine]-4,4'-dicarboxylate
SMILES [O-]C(=O)C1=CC=NC(=C1)C1=CC(=CC=N1)C([O-])=O

Specifications

Color White-Yellow
Quantity 1 g
Formula Weight 244.21
Percent Purity ≥96.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,2′-Bipyridine-4,4′-dicarboxylic Acid