missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,2'-Bipyrazine 97.0+%, TCI America™
$206.90 - $822.05
Chemical Identifiers
| CAS | 10199-00-5 |
|---|---|
| Molecular Formula | C8H6N4 |
| Molecular Weight (g/mol) | 158.164 |
| MDL Number | MFCD00010288 |
| InChI Key | DFXNVSIALRDJHY-UHFFFAOYSA-N |
| PubChem CID | 153669 |
| IUPAC Name | 2-pyrazin-2-ylpyrazine |
| SMILES | C1=CN=C(C=N1)C2=NC=CN=C2 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
B4297100MG
|
TCI America
B4297100MG |
100 mg |
Each for $206.90
|
|
|||||
|
B4297500MG
|
TCI America
B4297500MG |
500 mg |
Each for $822.05
|
|
|||||
Chemical Identifiers
| 10199-00-5 | |
| 158.164 | |
| DFXNVSIALRDJHY-UHFFFAOYSA-N | |
| 2-pyrazin-2-ylpyrazine |
| C8H6N4 | |
| MFCD00010288 | |
| 153669 | |
| C1=CN=C(C=N1)C2=NC=CN=C2 |
Specifications
| 10199-00-5 | |
| Yellow | |
| MFCD00010288 | |
| 2,2′C-Bipyrazinyl | |
| C1=CN=C(C=N1)C2=NC=CN=C2 | |
| 158.164 | |
| 158.16 | |
| Crystalline Powder |
| 188°C | |
| C8H6N4 | |
| 100 mg | |
| DFXNVSIALRDJHY-UHFFFAOYSA-N | |
| 2-pyrazin-2-ylpyrazine | |
| 153669 | |
| ≥97.0% (GC) | |
| 2,2′-Bipyrazine |
Safety and Handling
TSCA : No