missing translation for 'onlineSavingsMsg'
Learn More

2,2'-Bicinchoninic Acid 98.0+%, TCI America™

Catalog No. B35091G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B35091G 1 g
B35095G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B35091G Supplier TCI America Supplier No. B35091G
Only null left

Chemical Identifiers

CAS 1245-13-2
Molecular Formula C20H12N2O4
Molecular Weight (g/mol) 344.326
MDL Number MFCD00068342
InChI Key AFYNADDZULBEJA-UHFFFAOYSA-N
PubChem CID 71068
IUPAC Name 2-(4-carboxyquinolin-2-yl)quinoline-4-carboxylic acid
SMILES C1=CC=C2C(=C1)C(=CC(=N2)C3=NC4=CC=CC=C4C(=C3)C(=O)O)C(=O)O

Specifications

Melting Point 365°C
Quantity 1 g
Synonym 2,2′C-Biquinoline-4,4′C-dicarboxylic Acid
Formula Weight 344.33
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 2,2′-Bicinchoninic Acid