missing translation for 'onlineSavingsMsg'
Learn More

2,2'-Bi(1,8-naphthyridine), TCI America™

Catalog No. b1423100mg
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B1423100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B1423100MG Supplier TCI America Supplier No. B1423100MG

Chemical Identifiers

CAS 69110-33-4
Molecular Formula C16H10N4
Molecular Weight (g/mol) 258.284
MDL Number MFCD00142731
InChI Key OGSULPWGXWPZOJ-UHFFFAOYSA-N
PubChem CID 13185707
IUPAC Name 2-(1,8-naphthyridin-2-yl)-1,8-naphthyridine
SMILES C1=CC2=C(N=C1)N=C(C=C2)C3=NC4=C(C=CC=N4)C=C3

Specifications

Melting Point 227°C
Color Green-Yellow
Quantity 100 mg
Formula Weight 258.28
Physical Form Crystalline Powder
Chemical Name or Material 2,2′-Bi(1,8-naphthyridine)