missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,2', Azino-bis-(3-ethylbenzthiazoline sulfonic acid), diammonium salt, For TLC analysis, MP Biomedicals™
$445.69 - $445.69
Chemical Identifiers
| CAS | 30931-67-0 |
|---|---|
| Molecular Formula | C18H24N6O6S4 |
| Molecular Weight (g/mol) | 548.67 |
| MDL Number | MFCD00010404,MFCD00010404 |
| InChI Key | OHDRQQURAXLVGJ-AXMZSLBLSA-N |
| Synonym | ammonium 2,2'-hydrazine-1,2-diylidene bis 3-ethyl-2,3-dihydrobenzo d thiazole-6-sulfonate |
| PubChem CID | 91884754 |
| IUPAC Name | diazanium;3-ethyl-2-[(E)-(3-ethyl-6-sulfonato-1,3-benzothiazol-2-ylidene)hydrazinylidene]-1,3-benzothiazole-6-sulfonate |
| SMILES | [NH4+].[NH4+].CCN1\C(SC2=CC(=CC=C12)S([O-])(=O)=O)=N\N=C1/SC2=CC(=CC=C2N1CC)S([O-])(=O)=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
ICN19502305
|
MP Biomedicals Inc
0219502305 |
5 g |
Each for $445.69
|
|
|||||
Chemical Identifiers
| 30931-67-0 | |
| 548.67 | |
| OHDRQQURAXLVGJ-AXMZSLBLSA-N | |
| 91884754 | |
| [NH4+].[NH4+].CCN1\C(SC2=CC(=CC=C12)S([O-])(=O)=O)=N\N=C1/SC2=CC(=CC=C2N1CC)S([O-])(=O)=O |
| C18H24N6O6S4 | |
| MFCD00010404,MFCD00010404 | |
| ammonium 2,2'-hydrazine-1,2-diylidene bis 3-ethyl-2,3-dihydrobenzo d thiazole-6-sulfonate | |
| diazanium;3-ethyl-2-[(E)-(3-ethyl-6-sulfonato-1,3-benzothiazol-2-ylidene)hydrazinylidene]-1,3-benzothiazole-6-sulfonate |
Specifications
| 30931-67-0 | |
| C18H24N6O6S4 | |
| 5 g | |
| OHDRQQURAXLVGJ-AXMZSLBLSA-N | |
| diazanium;3-ethyl-2-[(E)-(3-ethyl-6-sulfonato-1,3-benzothiazol-2-ylidene)hydrazinylidene]-1,3-benzothiazole-6-sulfonate | |
| 91884754 | |
| Powder |
| Green | |
| MFCD00010404,MFCD00010404 | |
| ammonium 2,2'-hydrazine-1,2-diylidene bis 3-ethyl-2,3-dihydrobenzo d thiazole-6-sulfonate | |
| [NH4+].[NH4+].CCN1\C(SC2=CC(=CC=C12)S([O-])(=O)=O)=N\N=C1/SC2=CC(=CC=C2N1CC)S([O-])(=O)=O | |
| 548.67 | |
| 548.7 | |
| 2, 2' Azino-bis-(3-ethylbenzthiazoline sulfonic acid), diammonium salt, For TLC analysis |