missing translation for 'onlineSavingsMsg'
Learn More

2,2'-Azino-bis(3-ethylbenzothiazoline-6-sulfonic acid) diammonium salt, 98%

Catalog No. AAJ6553503
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6553503 1 g
AAJ6553506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6553503 Supplier Thermo Scientific Chemicals Supplier No. J6553503

Chromogenic substrate for peroxidase assays | CAS: 30931-67-0 | C18H24N6O6S4 | 548.67 g/mol

Chromogenic substrate for peroxidase assays.2,2'-Azino-bis(3-ethylbenzthiazoline-6-sulfonic acid) diammonium salt is widely utilized as a peroxidase substrate suitable for use in enzyme-linked immunosorbent assay (ELISA) procedures. It plays an important role in the food industry to measure the antioxidant capacities of foods. It is used to observe the reaction kinetics of specific enzymes and involved in glucose estimating reagent in blood serum.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Chromogenic substrate for peroxidase assays.2,2′-Azino-bis(3-ethylbenzthiazoline-6-sulfonic acid) diammonium salt is widely utilized as a peroxidase substrate suitable for use in enzyme-linked immunosorbent assay (ELISA) procedures. It plays an important role in the food industry to measure the antioxidant capacities of foods. It is used to observe the reaction kinetics of specific enzymes and involved in glucose estimating reagent in blood serum.

Solubility
Dissolve in water at 50mg/ml. Solution may be slightly hazy.

Notes
Light Sensitive. Keep container tightly closed in a dry and well-ventilated place.

Chemical Identifiers

CAS 30931-67-0
Molecular Formula C18H24N6O6S4
Molecular Weight (g/mol) 548.67
MDL Number MFCD00010404,MFCD00010404
InChI Key OHDRQQURAXLVGJ-AXMZSLBLSA-N
Synonym ammonium 2,2'-hydrazine-1,2-diylidene bis 3-ethyl-2,3-dihydrobenzo d thiazole-6-sulfonate
PubChem CID 91884754
IUPAC Name diazanium;3-ethyl-2-[(E)-(3-ethyl-6-sulfonato-1,3-benzothiazol-2-ylidene)hydrazinylidene]-1,3-benzothiazole-6-sulfonate
SMILES [NH4+].[NH4+].CCN1\C(SC2=CC(=CC=C12)S([O-])(=O)=O)=N\N=C1/SC2=CC(=CC=C2N1CC)S([O-])(=O)=O

Specifications

Color Green
Quantity 1 g
Beilstein 7329461
Sensitivity Light sensitive
Solubility Information Dissolve in water at 50mg/ml. Solution may be slightly hazy.
Formula Weight 548.68
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 2,2'-Azino-bis(3-ethylbenzothiazoline-6-sulfonic acid) diammonium salt
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 250-396-6
RTECSNumber DL7002000
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only