missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Aminophenyl)ethanol 95.0+%, TCI America™

Catalog No. p-7117840
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A23275G 5 g
A232725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A23275G Supplier TCI America Supplier No. A23275G
Only null left

Chemical Identifiers

CAS 5339-85-5
Molecular Formula C8H11NO
Molecular Weight (g/mol) 137.182
MDL Number MFCD00007754
InChI Key ILDXSRFKXABMHH-UHFFFAOYSA-N
Synonym 2-(2-Aminophenyl)ethyl Alcohol, 2-Aminophenethyl Alcohol
PubChem CID 79282
IUPAC Name 2-(2-aminophenyl)ethanol
SMILES C1=CC=C(C(=C1)CCO)N

Specifications

Color Yellow
Boiling Point 148°C
Quantity 5 g
Formula Weight 137.18
Percent Purity ≥95.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-(2-Aminophenyl)ethanol