missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Aminoethyl)thiophene 98.0+%, TCI America™

Catalog No. A163725G
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A163725G 25 g
A1637250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A163725G Supplier TCI America Supplier No. A163725G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 30433-91-1
Molecular Formula C6H9NS
Molecular Weight (g/mol) 127.205
MDL Number MFCD00051495
InChI Key HVLUYXIJZLDNIS-UHFFFAOYSA-N
Synonym thiophene-2-ethylamine, 2-thiopheneethylamine, 2-thiophen-2-yl ethanamine, thiopheneethanamine, 2-2-thienyl ethylamine, 2-thien-2-ylethanamine, 2-thiopheneethanamine, thiopheneethylamine, 2-thiophen-2-yl ethan-1-amine, 2-2-aminoethyl thiophene
PubChem CID 116521
IUPAC Name 2-thiophen-2-ylethanamine
SMILES C1=CSC(=C1)CCN

Specifications

Color Yellow
Boiling Point 201°C
Quantity 25 g
UN Number 2735
Formula Weight 127.21
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-(2-Aminoethyl)thiophene