missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Aminoethyl)pyridine, 98%

Catalog No. AAL0562703
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0562703 1 g
AAL0562706 5 g
AAL0562714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL0562703 Supplier Thermo Scientific Chemicals Supplier No. L0562703
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 2706-56-1 | C7H10N2 | 122.17 g/mol

2-(2-Aminoethyl)pyridine is used as active pharmaceutical ingredients.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(2-Aminoethyl)pyridine is used as active pharmaceutical ingredients.

Solubility
Soluble in water (partly), and most common polar solvent. Insoluble in non-polar solvents, and oils.

Notes
Air Sensitive. Store at 4°C. Stor in the dark. Incompatible with air, oxidizing agents, carbondioxide.

Chemical Identifiers

CAS 2706-56-1
Molecular Formula C7H10N2
Molecular Weight (g/mol) 122.17
MDL Number MFCD00006367
InChI Key XPQIPUZPSLAZDV-UHFFFAOYSA-N
Synonym 2-2-aminoethyl pyridine, 2-pyridylethylamine, 2-2-pyridyl ethylamine, 2-pyridin-2-yl ethanamine, 2-pyridineethanamine, demethylbetahistine, 2-pyridin-2-yl ethan-1-amine, alpha-pyridylethylamine, pyridine, 2-2-aminoethyl, 2-aminoethylpyridine
PubChem CID 75919
ChEBI CHEBI:74024
SMILES NCCC1=CC=CC=N1

Specifications

Density 1.02
Boiling Point 208°C
Flash Point 100°C (212°F)
Odor Amine-like
Refractive Index 1.536
Quantity 1 g
UN Number UN2735
Beilstein 111208
Sensitivity Air Sensitive
Solubility Information Soluble in water (partly),and most common polar solvent. Insoluble in non-polar solvents,and oils.
IUPAC Name 2-pyridin-2-ylethanamine
Formula Weight 122.17
Percent Purity 98%
Chemical Name or Material 2-(2-Aminoethyl)pyridine
Show More Show Less
Hazard Category H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
EINECSNumber 220-295-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only