missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Aminoethyl)-1-methylpyrrolidine 97.0+%, TCI America™

Catalog No. A23821G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A23821G 1 g
A23825G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A23821G Supplier TCI America Supplier No. A23821G
Only null left

Chemical Identifiers

CAS 51387-90-7
Molecular Formula C7H16N2
Molecular Weight (g/mol) 128.22
MDL Number MFCD00003175
InChI Key PNHGJPJOMCXSKN-UHFFFAOYNA-N
Synonym 2-2-aminoethyl-1-methylpyrrolidine, 2-1-methylpyrrolidin-2-yl ethanamine, 2-pyrrolidineethanamine, 1-methyl, 2-2'-aminoethyl-1-n-methylpyrrolidine, n-methyl-2-2-aminoethyl pyrrolidine, 1-methyl-2-2-aminoethyl pyrrolidine, 2-1-methylpyrrolidin-2-yl ethan-1-amine, 2-1-methyl-2-pyrrolidinyl ethylamine, 1-methyl-2-pyrrolidineethanamine, 1-methylpyrrolidine-2-ethylamine
PubChem CID 98388
IUPAC Name 2-(1-methylpyrrolidin-2-yl)ethan-1-amine
SMILES CN1CCCC1CCN

Specifications

Color Yellow
Quantity 1 g
UN Number 2735
Formula Weight 128.22
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-(2-Aminoethyl)-1-methylpyrrolidine