missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(2-Aminoethoxy)ethanol 98.0+%, TCI America™
$37.29 - $111.84
Chemical Identifiers
| CAS | 929-06-6 |
|---|---|
| Molecular Formula | C4H11NO2 |
| Molecular Weight (g/mol) | 105.137 |
| MDL Number | MFCD00008181 |
| InChI Key | GIAFURWZWWWBQT-UHFFFAOYSA-N |
| Synonym | 2-2-aminoethoxy ethanol, diglycolamine, 2-aminoethoxyethanol, ethanol, 2-2-aminoethoxy, diethylene glycol amine, 2-2-hydroxyethoxy ethylamine, diethylene glycol monoamine, 2-amino-2'-hydroxydiethyl ether, 1-amino-2-2-hydroxyethoxy ethane, 5-hydroxy-3-oxapentylamine |
| PubChem CID | 13578 |
| IUPAC Name | 2-(2-aminoethoxy)ethanol |
| SMILES | C(COCCO)N |
Chemical Identifiers
| 929-06-6 | |
| 105.137 | |
| GIAFURWZWWWBQT-UHFFFAOYSA-N | |
| 13578 | |
| C(COCCO)N |
| C4H11NO2 | |
| MFCD00008181 | |
| 2-2-aminoethoxy ethanol, diglycolamine, 2-aminoethoxyethanol, ethanol, 2-2-aminoethoxy, diethylene glycol amine, 2-2-hydroxyethoxy ethylamine, diethylene glycol monoamine, 2-amino-2'-hydroxydiethyl ether, 1-amino-2-2-hydroxyethoxy ethane, 5-hydroxy-3-oxapentylamine | |
| 2-(2-aminoethoxy)ethanol |
Specifications
| 929-06-6 | |
| 10.2 (10g/L 20°C) | |
| C4H11NO2 | |
| 25 mL | |
| 2-2-aminoethoxy ethanol, diglycolamine, 2-aminoethoxyethanol, ethanol, 2-2-aminoethoxy, diethylene glycol amine, 2-2-hydroxyethoxy ethylamine, diethylene glycol monoamine, 2-amino-2'-hydroxydiethyl ether, 1-amino-2-2-hydroxyethoxy ethane, 5-hydroxy-3-oxapentylamine | |
| C(COCCO)N | |
| 105.137 | |
| 105.14 | |
| Liquid |
| Yellow | |
| 223°C | |
| MFCD00008181 | |
| 3055 | |
| GIAFURWZWWWBQT-UHFFFAOYSA-N | |
| 2-(2-aminoethoxy)ethanol | |
| 13578 | |
| ≥98.0% (GC,T) | |
| 2-(2-Aminoethoxy)ethanol |
Safety and Handling
EINECSNumber : (2)-3202
RTECSNumber : KJ6125000
TSCA : Yes