missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Aminoethoxy)ethanol 98.0+%, TCI America™

Catalog No. A0301500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A0301500ML 500 mL
A030125ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A0301500ML Supplier TCI America Supplier No. A0301500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 929-06-6
Molecular Formula C4H11NO2
Molecular Weight (g/mol) 105.137
MDL Number MFCD00008181
InChI Key GIAFURWZWWWBQT-UHFFFAOYSA-N
Synonym 2-2-aminoethoxy ethanol, diglycolamine, 2-aminoethoxyethanol, ethanol, 2-2-aminoethoxy, diethylene glycol amine, 2-2-hydroxyethoxy ethylamine, diethylene glycol monoamine, 2-amino-2'-hydroxydiethyl ether, 1-amino-2-2-hydroxyethoxy ethane, 5-hydroxy-3-oxapentylamine
PubChem CID 13578
IUPAC Name 2-(2-aminoethoxy)ethanol
SMILES C(COCCO)N

Specifications

Color Yellow
pH 10.2 (10g/L 20°C)
Boiling Point 223°C
Quantity 500 mL
UN Number 3055
Formula Weight 105.14
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-(2-Aminoethoxy)ethanol
EINECSNumber (2)-3202
RTECSNumber KJ6125000
TSCA Yes