Learn More
2-(2-Aminoethoxy)ethanol, 98%
CAS: 929-06-6 | C4H11NO2 | 105.14 g/mol
Supplier: Thermo Scientific Chemicals 103630010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 2-(2-Aminoethoxy)ethanol | |
| -12.5°C | |
| 1.0600g/mL | |
| 127°C | |
| 97.5% min. (GC) | |
| C4H11NO2 | |
| H2NCH2CH2OCH2CH2OH | |
| 04,III,642 | |
| 2-2-aminoethoxy ethanol, diglycolamine, 2-aminoethoxyethanol, ethanol, 2-2-aminoethoxy, diethylene glycol amine, 2-2-hydroxyethoxy ethylamine, diethylene glycol monoamine, 2-amino-2'-hydroxydiethyl ether, 1-amino-2-2-hydroxyethoxy ethane, 5-hydroxy-3-oxapentylamine | |
| GIAFURWZWWWBQT-UHFFFAOYSA-N | |
| 2-(2-aminoethoxy)ethanol | |
| 13578 | |
| 98% |
| 929-06-6 | |
| Yellow | |
| 218.0°C to 224.0°C | |
| Authentic | |
| Glass bottle | |
| 1.4585 to 1.4605 | |
| 1 L | |
| 1.06 | |
| Solubility in water: miscible. Other solubilities: miscible with alcohol and aromatic hydrocarbons, practically immiscible with aliphatic hydrocarbons, and ethyl ether | |
| C(COCCO)N | |
| 105.14 | |
| 105.14 | |
| Crystalline Powder and/or Chunks |
Chemical Identifiers
| 929-06-6 | |
| 105.14 | |
| 2-2-aminoethoxy ethanol, diglycolamine, 2-aminoethoxyethanol, ethanol, 2-2-aminoethoxy, diethylene glycol amine, 2-2-hydroxyethoxy ethylamine, diethylene glycol monoamine, 2-amino-2'-hydroxydiethyl ether, 1-amino-2-2-hydroxyethoxy ethane, 5-hydroxy-3-oxapentylamine | |
| 2-(2-aminoethoxy)ethanol |
| C4H11NO2 | |
| GIAFURWZWWWBQT-UHFFFAOYSA-N | |
| 13578 | |
| C(COCCO)N |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat
GHS Signal Word: Danger
EINECSNumber : 213-195-4
RUO – Research Use Only