missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Aminoethoxy)ethanol, 98%

Catalog No. AAL1889722
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1889722 100 g
AAL1889736 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1889722 Supplier Thermo Scientific Chemicals Supplier No. L1889722
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 929-06-6 | C4H11NO2 | 105.137 g/mol

2-(2-Aminoethoxy)ethanol is used mainly in stripper solutions for applications in electronics, in gas treating to remove carbon dioxide and hydrogen sulfide, and in coolants/lubricants for metal working applications. Other applications of aminoethoxyethanol (ADG) include, inter alia, crop protection products, surfactants and colorants. It is also used as a selective solvent for recovery of aromatics from refinery streams and in preparation of foam stabilizers, wetting and emulsifying agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(2-Aminoethoxy)ethanol is used mainly in stripper solutions for applications in electronics, in gas treating to remove carbon dioxide and hydrogen sulfide, and in coolants/lubricants for metal working applications. Other applications of aminoethoxyethanol (ADG) include, inter alia, crop protection products, surfactants and colorants. It is also used as a selective solvent for recovery of aromatics from refinery streams and in preparation of foam stabilizers, wetting and emulsifying agents.

Solubility
Fully miscible in water, alcohols and aromatic hydrocarbons.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. It is sensitive to air. Store under inert gas.

Chemical Identifiers

CAS 929-06-6
Molecular Formula C4H11NO2
Molecular Weight (g/mol) 105.137
MDL Number MFCD00008181
InChI Key GIAFURWZWWWBQT-UHFFFAOYSA-N
Synonym 2-2-aminoethoxy ethanol, diglycolamine, 2-aminoethoxyethanol, ethanol, 2-2-aminoethoxy, diethylene glycol amine, 2-2-hydroxyethoxy ethylamine, diethylene glycol monoamine, 2-amino-2'-hydroxydiethyl ether, 1-amino-2-2-hydroxyethoxy ethane, 5-hydroxy-3-oxapentylamine
PubChem CID 13578
IUPAC Name 2-(2-aminoethoxy)ethanol
SMILES C(COCCO)N

Specifications

Melting Point -11°C
Density 1.057
Boiling Point 125°C to 131°C (23 mmHg)
Flash Point 127°C (260°F)
Refractive Index 1.457
Quantity 100 g
UN Number UN3055
Beilstein 906728
Sensitivity Air Sensitive
Solubility Information Fully miscible in water,alcohols and aromatic hydrocarbons.
Formula Weight 105.14
Percent Purity 98%
Chemical Name or Material 2-(2-Aminoethoxy)ethanol
Show More Show Less
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318-H312
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful in contact with skin.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: 2-(2-AMINOETHOXY)ETHANOL
EINECSNumber 213-195-4
RTECSNumber KJ6125000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only