missing translation for 'onlineSavingsMsg'
Learn More

2,2,7-Trimethyloctane-3,5-dione, 97%

Catalog No. AAL1463903
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1463903 1 g
AAL1463906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1463903 Supplier Thermo Scientific Chemicals Supplier No. L1463903

CAS: 69725-37-7 | C11H20O2 | 184.279 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 69725-37-7
Molecular Formula C11H20O2
Molecular Weight (g/mol) 184.279
MDL Number MFCD00511279
InChI Key KEBPOWLGOOTMPK-UHFFFAOYSA-N
PubChem CID 112295
IUPAC Name 2,2,7-trimethyloctane-3,5-dione
SMILES CC(C)CC(=O)CC(=O)C(C)(C)C

Specifications

Boiling Point 44°C (0.2 mmHg)
Refractive Index 1.458
Quantity 1 g
Synonym 2,2,7-trimethyl-3,5-octanedione, 3,5-octanedione, 2,2,7-trimethyl, 2,2 7-trimethyloctane-3,5-dione, 3,5-octanedione,2,2,7-trimethyl, 2,2,7-trimethyl-3,5-octanedione #, 2,2,7-trimethyl-3,5-octadione
Formula Weight 184.28
Percent Purity ≥98%
Chemical Name or Material 2,2,7-Trimethyloctane-3,5-dione
EINECSNumber 000-000-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only