missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,2,6-Trimethylcyclohexanone, 97%
Description
2,2,6-Trimethylcyclohexanone is used as a flavoring agent. It is also used to prepare terpene viz. beta-ionone.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2,2,6-Trimethylcyclohexanone is used as a flavoring agent. It is also used to prepare terpene viz. beta-ionone.
Solubility
Slightly miscible with water.
Notes
Incompatible with strong oxidizing agents and strong reducing agents.
2,2,6-Trimethylcyclohexanone is used as a flavoring agent. It is also used to prepare terpene viz. beta-ionone.
Solubility
Slightly miscible with water.
Notes
Incompatible with strong oxidizing agents and strong reducing agents.
Specifications
Specifications
| Density | 0.904 |
| Boiling Point | 178°C to 179°C |
| Flash Point | 51°C (125°F) |
| Refractive Index | 1.447 |
| Quantity | 1 g |
| UN Number | UN1224 |
| Beilstein | 1905593 |
| Solubility Information | Slightly miscible with water. |
| Formula Weight | 140.23 |
| Percent Purity | 97% |
| Show More |
Safety and Handling
| missing translation for 'healthHazard1' | H226 |
| missing translation for 'healthHazard2' | GHS H Statement H226-H315-H319-H335 Flammable liquid and vapour. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| missing translation for 'healthHazard3' | P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: KETONES, LIQUID, N.O.S. |
| EINECSNumber | 219-309-9 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only