missing translation for 'onlineSavingsMsg'
Learn More

2,2,6-Trimethyl-4H-1,3-dioxin-4-one, 94%

Catalog No. AC296911000
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC296911000 100 g Glass bottle
AC296915000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC296911000 Supplier Thermo Scientific Chemicals Supplier No. 296911000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 5394-63-8 | C7H10O3 | 142.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5394-63-8
Molecular Formula C7H10O3
Molecular Weight (g/mol) 142.15
MDL Number MFCD00040468
InChI Key XFRBXZCBOYNMJP-UHFFFAOYSA-N
Synonym 2,2,6-trimethyl-4h-1,3-dioxin-4-one, 2,2,4-trimethyl-6-keto-1,3-dioxin, diketene acetone adduct, 4h-1,3-dioxin-4-one, 2,2,6-trimethyl, 2,2,6-trimethyl-1,3-dioxen-4-one, 2,2,6-trimethyl-1,3-dioxine-4-one, 2,2,6-trimethyl-1,3 dioxin-4-one, pubchem20364, 2,2,6-trimethyl-4h-1,3-dioxin-4-on, acmc-1akcb
PubChem CID 79368
IUPAC Name 2,2,6-trimethyl-1,3-dioxin-4-one
SMILES CC1=CC(=O)OC(O1)(C)C

Specifications

CAS Min % 90.0
CAS Max % 100.0
Melting Point 4°C to 13°C
Color Brown
Density 1.0800g/mL
Boiling Point 65°C to 67°C (2.7 mbar)
Flash Point 14°C
Infrared Spectrum Authentic
Assay Percent Range 92% min. (GC)
Packaging Glass bottle
Refractive Index 1.4590 to 1.462
Quantity 100 g
Beilstein 19, IV, 1604
Specific Gravity 1.08
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in acetone and methanol
Formula Weight 142.15
Percent Purity 94%
Physical Form Liquid
Chemical Name or Material 2,2,6-Trimethyl-4H-1,3-dioxin-4-one
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
EINECSNumber 226-403-3
Recommended Storage product darkens with time

RUO – Research Use Only