missing translation for 'onlineSavingsMsg'
Learn More

2,2,6-Trimethyl-1,3-dioxin-4-one, cont. up to ca 6% acetone

Catalog No. AAA1425414
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1425414 25 g
AAA1425422 100 g
AAA1425436 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog No. AAA1425414 Supplier Thermo Scientific Chemicals Supplier No. A1425414

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.

CAS: 5394-63-8 | C7H10O3 | 142.154 g/mol

2,2,6-Trimethyl-1,3-dioxin-4-one is used in the synthesis of dihydropyrimininones as picomolar sodium iodide symporter inhibitors. It is also used in the synthesis of resorcinol amide Hsp90 Inhibitor AT13387. It is used in the synthesis of acetylketene by flash pyrolysis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2,6-Trimethyl-1,3-dioxin-4-one is used in the synthesis of dihydropyrimininones as picomolar sodium iodide symporter inhibitors. It is also used in the synthesis of resorcinol amide Hsp90 Inhibitor AT13387. It is used in the synthesis of acetylketene by flash pyrolysis.

Solubility
Soluble in Chloroform, Methanol. Not miscible in water.

Notes
Store at 4°C. Protect from heat. Store away from oxidizing agents.

Chemical Identifiers

CAS 5394-63-8
Molecular Formula C7H10O3
Molecular Weight (g/mol) 142.154
MDL Number MFCD00040468
InChI Key XFRBXZCBOYNMJP-UHFFFAOYSA-N
Synonym 2,2,6-trimethyl-4h-1,3-dioxin-4-one, 2,2,4-trimethyl-6-keto-1,3-dioxin, diketene acetone adduct, 4h-1,3-dioxin-4-one, 2,2,6-trimethyl, 2,2,6-trimethyl-1,3-dioxen-4-one, 2,2,6-trimethyl-1,3-dioxine-4-one, 2,2,6-trimethyl-1,3 dioxin-4-one, pubchem20364, 2,2,6-trimethyl-4h-1,3-dioxin-4-on, acmc-1akcb
PubChem CID 79368
IUPAC Name 2,2,6-trimethyl-1,3-dioxin-4-one
SMILES CC1=CC(=O)OC(O1)(C)C

Specifications

Melting Point 7°C
Density 1.094
Boiling Point 65°C to 67°C (2 mmHg)
Flash Point 14°C (57°F)
Refractive Index 1.461
Quantity 25 g
UN Number UN1993
Beilstein 2408
Solubility Information Soluble in Chloroform,Methanol. Not miscible in water.
Formula Weight 142.15
Percent Purity 94%
Assay cont. up to ≈6% acetone
Chemical Name or Material 2,2,6-Trimethyl-1,3-dioxin-4-one
Show More Show Less
Hazard Category H225-H319-H336
Hazard Statement GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P240-P241-P242-P243-P264b-P280-P303+P361+P353-P305+P351+P338-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 226-403-3
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only