missing translation for 'onlineSavingsMsg'
Learn More

2-(2,6-Dichlorophenyl)ethanol 97.0+%, TCI America™

Catalog No. D29225G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D29225G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D29225G Supplier TCI America Supplier No. D29225G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 30595-79-0
Molecular Formula C8H8Cl2O
Molecular Weight (g/mol) 191.05
MDL Number MFCD00800674
InChI Key ZBQPKQUIKJDGIX-UHFFFAOYSA-N
Synonym 2,6-dichlorophenethyl alcohol, 2-2,6-dichlorophenyl ethanol, 2,6-dichlorophenethylalcohol, 2-2,6-dichlorophenyl ethan-1-ol, 2,6-dichlorophenethanol, benzeneethanol, 2,6-dichloro, acmc-209hha, 2,6-dichlorobenzeneethanol, ksc493m0h, benzeneethanol,2,6-dichloro
PubChem CID 2734100
IUPAC Name 2-(2,6-dichlorophenyl)ethan-1-ol
SMILES OCCC1=C(Cl)C=CC=C1Cl

Specifications

Melting Point 83°C
Color White-Yellow
Boiling Point 152°C
Quantity 5 g
Formula Weight 191.05
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-(2,6-Dichlorophenyl)ethanol