missing translation for 'onlineSavingsMsg'
Learn More

2,2,6,6-Tetramethyl-3,6-dihydro-2H-pyran-4-boronic acid pinacol ester, 97%

Catalog No. AC445530010
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC445530010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC445530010 Supplier Thermo Scientific Chemicals Supplier No. 445530010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1142363-56-1 | C15H27BO3 | 266.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1142363-56-1
Molecular Formula C15H27BO3
Molecular Weight (g/mol) 266.19
MDL Number MFCD11044439
InChI Key VOXXPMAJNSVUKJ-UHFFFAOYSA-N
Synonym 2,2,6,6-tetramethyl-3,6-dihydro-2h-pyran-4-yl boronic acid, pinacol ester, 4,4,5,5-tetramethyl-2-2,2,6,6-tetramethyl-3,6-dihydro-2h-pyran-4-yl-1,3,2-dioxaborolane, 2,2,6,6-tetramethyl-3,6-dihydro-2h-pyran-4-boronic acid pinacol ester, 2,2,6,6-tetramethyl-3,6-dihydro-2h-pyran-4-yl boronic acid pinacol ester, 2,2,6,6-tetramethyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-3,6-dihydro-2h-pyran, 4,4,5,5-tetramethyl-2-2,2,6,6-tetramethyl-3h-pyran-4-yl-1,3,2-dioxaborolane, 2,2,6,6-tetramethyl-3,6-dihydro-2h-pyran-4-yl bor, 2,2,6,6-tetramethyl-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-3,6-dihydro-2h-pyran
PubChem CID 46738000
IUPAC Name 4,4,5,5-tetramethyl-2-(2,2,6,6-tetramethyl-3H-pyran-4-yl)-1,3,2-dioxaborolane
SMILES CC1(C)OB(OC1(C)C)C1=CC(C)(C)OC(C)(C)C1

Specifications

Melting Point 91°C to 93°C
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 266.19
Percent Purity 97%
Chemical Name or Material 2,2,6,6-Tetramethyl-3,6-dihydro-2H-pyran-4-boronic acid pinacol ester
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for

RUO – Research Use Only