missing translation for 'onlineSavingsMsg'
Learn More

2,2,5,5-Tetramethyl-3-pyrrolin-1-oxyl-3-carboxylic acid, 99%

Catalog No. ac420640010
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC420640010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC420640010 Supplier Thermo Scientific Chemicals Supplier No. 420640010

CAS: 2154-67-8 | C9H14NO3 | 184.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2154-67-8
Molecular Formula C9H14NO3
Molecular Weight (g/mol) 184.22
MDL Number MFCD00041847
Synonym 3-Carboxy-2,2,5,5-tetramethyl-3-pyrrolin-1-yloxy

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 202°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Quantity 1 g
Formula Weight 184.22
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 2,2,5,5-Tetramethyl-3-pyrrolin-1-oxyl-3-carboxylic acid, 99%
Show More Show Less
EINECSNumber 218-447-7
TSCA TSCA

RUO – Research Use Only