missing translation for 'onlineSavingsMsg'
Learn More

2,2':5',2″-Terthiophene-5-carboxaldehyde 98.0+%, TCI America™

Catalog No. T18051G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T18051G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T18051G Supplier TCI America Supplier No. T18051G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7342-41-8
Molecular Formula C13H8OS3
Molecular Weight (g/mol) 276.39
MDL Number MFCD00115181
InChI Key PMPDDPJYARBNGV-UHFFFAOYSA-N
Synonym 2,2':5',2-terthiophene-5-carboxaldehyde, 5-5-2-thienyl-2-thienyl thiophene-2-carbaldehyde, 2,2':5',2-terthiophene-5-carbaldehyde, 5-formyl-2,2':5',2-terthiophene, chembl91933, 5-5-thiophen-2-ylthiophen-2-yl thiophene-2-carbaldehyde, 5-5-thiophen-2-yl thiophen-2-yl thiophene-2-carbaldehyde, 2-formyl-alpha-terthienyl
PubChem CID 454742
IUPAC Name 5'-(thiophen-2-yl)-[2,2'-bithiophene]-5-carbaldehyde
SMILES O=CC1=CC=C(S1)C1=CC=C(S1)C1=CC=CS1

Specifications

Melting Point 141°C
Quantity 1 g
Formula Weight 276.39
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 2,2′:5′,2′′-Terthiophene-5-carboxaldehyde