missing translation for 'onlineSavingsMsg'
Learn More

2,2,4-Trimethylhexane 98.0+%, TCI America™

Catalog No. T07745ML
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T07745ML 5 mL
T07741ML 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T07745ML Supplier TCI America Supplier No. T07745ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 16747-26-5
Molecular Formula C9H20
Molecular Weight (g/mol) 128.26
MDL Number MFCD00039928
InChI Key AFTPEBDOGXRMNQ-UHFFFAOYNA-N
PubChem CID 28022
IUPAC Name 2,2,4-trimethylhexane
SMILES CCC(C)CC(C)(C)C

Specifications

Color Colorless
Boiling Point 126°C
Quantity 5 mL
UN Number 1920
Formula Weight 128.26
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,2,4-Trimethylhexane
EINECSNumber (2)-0009
TSCA No