missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,2,4-Trimethyl-1,3-pentanediol 97.0+%, TCI America™
$54.06 - $281.48
Chemical Identifiers
| CAS | 144-19-4 |
|---|---|
| Molecular Formula | C8H18O2 |
| Molecular Weight (g/mol) | 146.23 |
| MDL Number | MFCD00004681 |
| InChI Key | JCTXKRPTIMZBJT-UHFFFAOYNA-N |
| Synonym | 2,2,4-trimethyl-1,3-pentanediol, tmpd, 1,3-pentanediol, 2,2,4-trimethyl, tmpd alcohol, caswell no. 893, tmpd glycol, epa pesticide chemical code 041002, 2,2,4-trimethyl pentanediol-1,3, trimethyl-1,3-pentanediol, acmc-1brgn |
| PubChem CID | 8946 |
| IUPAC Name | 2,2,4-trimethylpentane-1,3-diol |
| SMILES | CC(C)C(O)C(C)(C)CO |
Chemical Identifiers
| 144-19-4 | |
| 146.23 | |
| JCTXKRPTIMZBJT-UHFFFAOYNA-N | |
| 8946 | |
| CC(C)C(O)C(C)(C)CO |
| C8H18O2 | |
| MFCD00004681 | |
| 2,2,4-trimethyl-1,3-pentanediol, tmpd, 1,3-pentanediol, 2,2,4-trimethyl, tmpd alcohol, caswell no. 893, tmpd glycol, epa pesticide chemical code 041002, 2,2,4-trimethyl pentanediol-1,3, trimethyl-1,3-pentanediol, acmc-1brgn | |
| 2,2,4-trimethylpentane-1,3-diol |
Specifications
| 144-19-4 | |
| White | |
| C8H18O2 | |
| 25 g | |
| JCTXKRPTIMZBJT-UHFFFAOYNA-N | |
| 2,2,4-trimethylpentane-1,3-diol | |
| 8946 | |
| ≥97.0% (GC) | |
| 2,2,4-Trimethyl-1,3-pentanediol |
| 54°C | |
| 232°C | |
| MFCD00004681 | |
| 2,2,4-trimethyl-1,3-pentanediol, tmpd, 1,3-pentanediol, 2,2,4-trimethyl, tmpd alcohol, caswell no. 893, tmpd glycol, epa pesticide chemical code 041002, 2,2,4-trimethyl pentanediol-1,3, trimethyl-1,3-pentanediol, acmc-1brgn | |
| CC(C)C(O)C(C)(C)CO | |
| 146.23 | |
| 146.23 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (2)-0240
RTECSNumber : SA1400000
TSCA : Yes