missing translation for 'onlineSavingsMsg'
Learn More

2,2,4-Trimethyl-1,3-pentanediol Diisobutyrate 98.5+%, TCI America™

Catalog No. T099725ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T099725ML 25 mL
T0997500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T099725ML Supplier TCI America Supplier No. T099725ML

Chemical Identifiers

CAS 6846-50-0
Molecular Formula C16H30O4
Molecular Weight (g/mol) 286.412
MDL Number MFCD00059267
InChI Key OMVSWZDEEGIJJI-UHFFFAOYSA-N
Synonym 2,2,4-trimethyl-1,3-pentanediol diisobutyrate, kodaflex txib, 2,2,4-trimethylpentane-1,3-diyl bis 2-methylpropanoate, txib, propanoic acid, 2-methyl-, 2,2-dimethyl-1-1-methylethyl-1,3-propanediyl ester, 2,2,4-trimethylpentanediol-1,3-diisobutyrate, isobutyric acid, 1-isopropyl-2,2-dimethyltrimethylene ester, 1-isopropyl-2,2-dimethyltrimethylene diisobutyrate, 2,2,4-trimethyl-1,3-pentanediol ester, dsstox_cid_7635
PubChem CID 23284
IUPAC Name [2,2,4-trimethyl-3-(2-methylpropanoyloxy)pentyl] 2-methylpropanoate
SMILES CC(C)C(C(C)(C)COC(=O)C(C)C)OC(=O)C(C)C

Specifications

Melting Point -70°C
Color Colorless
Boiling Point 280°C
Quantity 25 mL
Formula Weight 286.41
Percent Purity ≥98.5% (GC)
Physical Form Liquid
Chemical Name or Material 2,2,4-Trimethyl-1,3-pentanediol Diisobutyrate
EINECSNumber (2)-2498
RTECSNumber SA1420000
TSCA Yes