missing translation for 'onlineSavingsMsg'
Learn More

2,2,4-Trimethyl-1,3-dioxolane 98.0+%, TCI America™

Catalog No. T257525G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T257525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T257525G Supplier TCI America Supplier No. T257525G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1193-11-9
Molecular Formula C6H12O2
Molecular Weight (g/mol) 116.16
MDL Number MFCD00090841
InChI Key ALTFLAPROMVXNX-UHFFFAOYNA-N
Synonym propylene glycol acetone ketal, 1,3-dioxolane, 2,2,4-trimethyl, acetone propylene glycol acetal, 1,3-dioxolane,2,2,4-trimethyl, fema no. 3441, 2,2,4-trimethyl-1,3-oxacyclopentane, 2,2,4-trimethyl-1,3-dioxacyclopentane, propylene glycol acetonide, 1,2-propanediol acetonide, 2,2,4-trimethyldioxolane
PubChem CID 62384
IUPAC Name 2,2,4-trimethyl-1,3-dioxolane
SMILES CC1COC(C)(C)O1

Specifications

Color Yellow
Boiling Point 98°C
Quantity 25 g
UN Number 1993
Formula Weight 116.16
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,2,4-Trimethyl-1,3-dioxolane