missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,2',4'-Trichloroacetophenone 98.0+%, TCI America™
$74.56 - $415.68
Chemical Identifiers
| CAS | 4252-78-2 |
|---|---|
| Molecular Formula | C8H5Cl3O |
| Molecular Weight (g/mol) | 223.477 |
| MDL Number | MFCD00000934 |
| InChI Key | VYWPPRLJNVHPEU-UHFFFAOYSA-N |
| Synonym | 2,2',4'-trichloroacetophenone, 2-chloro-1-2,4-dichlorophenyl ethanone, 2,4-dichlorophenacyl chloride, unii-2e35p7hji2, 2-chloro-1-2,4-dichlorophenyl ethan-1-one, ccris 633, ethanone, 2-chloro-1-2,4-dichlorophenyl, 2,2,4-trichloroacetophenone, dsstox_cid_6190, 1-2,4-dichlorophenyl-2-chloroethan-1-one |
| PubChem CID | 20250 |
| IUPAC Name | 2-chloro-1-(2,4-dichlorophenyl)ethanone |
| SMILES | C1=CC(=C(C=C1Cl)Cl)C(=O)CCl |
Chemical Identifiers
| 4252-78-2 | |
| 223.477 | |
| VYWPPRLJNVHPEU-UHFFFAOYSA-N | |
| 20250 | |
| C1=CC(=C(C=C1Cl)Cl)C(=O)CCl |
| C8H5Cl3O | |
| MFCD00000934 | |
| 2,2',4'-trichloroacetophenone, 2-chloro-1-2,4-dichlorophenyl ethanone, 2,4-dichlorophenacyl chloride, unii-2e35p7hji2, 2-chloro-1-2,4-dichlorophenyl ethan-1-one, ccris 633, ethanone, 2-chloro-1-2,4-dichlorophenyl, 2,2,4-trichloroacetophenone, dsstox_cid_6190, 1-2,4-dichlorophenyl-2-chloroethan-1-one | |
| 2-chloro-1-(2,4-dichlorophenyl)ethanone |
Specifications
| 4252-78-2 | |
| C8H5Cl3O | |
| 25 g | |
| 2,2',4'-trichloroacetophenone, 2-chloro-1-2,4-dichlorophenyl ethanone, 2,4-dichlorophenacyl chloride, unii-2e35p7hji2, 2-chloro-1-2,4-dichlorophenyl ethan-1-one, ccris 633, ethanone, 2-chloro-1-2,4-dichlorophenyl, 2,2,4-trichloroacetophenone, dsstox_cid_6190, 1-2,4-dichlorophenyl-2-chloroethan-1-one | |
| C1=CC(=C(C=C1Cl)Cl)C(=O)CCl | |
| 223.477 | |
| 223.48 | |
| Crystalline Powder |
| White | |
| MFCD00000934 | |
| 1759 | |
| VYWPPRLJNVHPEU-UHFFFAOYSA-N | |
| 2-chloro-1-(2,4-dichlorophenyl)ethanone | |
| 20250 | |
| ≥98.0% (GC) | |
| 2,2′,4′-Trichloroacetophenone |
Safety and Handling
TSCA : No