Learn More
2,2',4'-Trichloroacetophenone, 97%
CAS: 4252-78-2 | C8H5Cl3O | 223.477 g/mol
$181.82 - $693.77
Chemical Identifiers
| CAS | 4252-78-2 |
|---|---|
| Molecular Formula | C8H5Cl3O |
| Molecular Weight (g/mol) | 223.477 |
| MDL Number | MFCD00000934 |
| InChI Key | VYWPPRLJNVHPEU-UHFFFAOYSA-N |
| Synonym | 2,2',4'-trichloroacetophenone, 2-chloro-1-2,4-dichlorophenyl ethanone, 2,4-dichlorophenacyl chloride, unii-2e35p7hji2, 2-chloro-1-2,4-dichlorophenyl ethan-1-one, ccris 633, ethanone, 2-chloro-1-2,4-dichlorophenyl, 2,2,4-trichloroacetophenone, dsstox_cid_6190, 1-2,4-dichlorophenyl-2-chloroethan-1-one |
| PubChem CID | 20250 |
| IUPAC Name | 2-chloro-1-(2,4-dichlorophenyl)ethanone |
| SMILES | C1=CC(=C(C=C1Cl)Cl)C(=O)CCl |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1083522
|
Thermo Scientific Chemicals
A1083522 |
100 g |
Each for $181.82
|
|
|||||
|
AAA1083536
|
Thermo Scientific Chemicals
A1083536 |
500 g |
Each for $693.77
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 4252-78-2 | |
| 223.477 | |
| VYWPPRLJNVHPEU-UHFFFAOYSA-N | |
| 20250 | |
| C1=CC(=C(C=C1Cl)Cl)C(=O)CCl |
| C8H5Cl3O | |
| MFCD00000934 | |
| 2,2',4'-trichloroacetophenone, 2-chloro-1-2,4-dichlorophenyl ethanone, 2,4-dichlorophenacyl chloride, unii-2e35p7hji2, 2-chloro-1-2,4-dichlorophenyl ethan-1-one, ccris 633, ethanone, 2-chloro-1-2,4-dichlorophenyl, 2,2,4-trichloroacetophenone, dsstox_cid_6190, 1-2,4-dichlorophenyl-2-chloroethan-1-one | |
| 2-chloro-1-(2,4-dichlorophenyl)ethanone |
Specifications
| 4252-78-2 | |
| >110°C (230°F) | |
| MFCD00000934 | |
| UN2923 | |
| 2,2',4'-trichloroacetophenone, 2-chloro-1-2,4-dichlorophenyl ethanone, 2,4-dichlorophenacyl chloride, unii-2e35p7hji2, 2-chloro-1-2,4-dichlorophenyl ethan-1-one, ccris 633, ethanone, 2-chloro-1-2,4-dichlorophenyl, 2,2,4-trichloroacetophenone, dsstox_cid_6190, 1-2,4-dichlorophenyl-2-chloroethan-1-one | |
| C1=CC(=C(C=C1Cl)Cl)C(=O)CCl | |
| 223.477 | |
| 223.49 | |
| 2,2',4'-Trichloroacetophenone |
| 52°C to 56°C | |
| C8H5Cl3O | |
| 100 g | |
| 957098 | |
| VYWPPRLJNVHPEU-UHFFFAOYSA-N | |
| 2-chloro-1-(2,4-dichlorophenyl)ethanone | |
| 20250 | |
| 97% |
Safety and Handling
GHS H Statement
H301-H314-H318-H335
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause respiratory irritation.
P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H301-H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S.
EINECSNumber : 224-218-2
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only