missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(2,4-Dihydroxyphenyl)-4,6-diphenyl-1,3,5-triazine 98.0+%, TCI America™
$186.41 - $553.62
Chemical Identifiers
| CAS | 38369-95-8 |
|---|---|
| Molecular Formula | C21H15N3O2 |
| Molecular Weight (g/mol) | 341.37 |
| MDL Number | MFCD16251126 |
| InChI Key | JUMDJIDYKLZVAF-UHFFFAOYSA-N |
| PubChem CID | 53630481 |
| IUPAC Name | 4-(4,6-diphenyl-1H-1,3,5-triazin-2-ylidene)-3-hydroxycyclohexa-2,5-dien-1-one |
| SMILES | C1=CC=C(C=C1)C2=NC(=NC(=C3C=CC(=O)C=C3O)N2)C4=CC=CC=C4 |
Chemical Identifiers
| 38369-95-8 | |
| 341.37 | |
| JUMDJIDYKLZVAF-UHFFFAOYSA-N | |
| 4-(4,6-diphenyl-1H-1,3,5-triazin-2-ylidene)-3-hydroxycyclohexa-2,5-dien-1-one |
| C21H15N3O2 | |
| MFCD16251126 | |
| 53630481 | |
| C1=CC=C(C=C1)C2=NC(=NC(=C3C=CC(=O)C=C3O)N2)C4=CC=CC=C4 |
Specifications
| 38369-95-8 | |
| Yellow | |
| MFCD16251126 | |
| JUMDJIDYKLZVAF-UHFFFAOYSA-N | |
| 4-(4,6-diphenyl-1H-1,3,5-triazin-2-ylidene)-3-hydroxycyclohexa-2,5-dien-1-one | |
| 53630481 | |
| ≥98.0% (HPLC,N) | |
| 2-(2,4-Dihydroxyphenyl)-4,6-diphenyl-1,3,5-triazine |
| 276°C | |
| C21H15N3O2 | |
| 1 g | |
| C1=CC=C(C=C1)C2=NC(=NC(=C3C=CC(=O)C=C3O)N2)C4=CC=CC=C4 | |
| 341.37 | |
| 341.37 | |
| Crystalline Powder |
Safety and Handling
TSCA : No