missing translation for 'onlineSavingsMsg'
Learn More

2-(2,4-Dichlorophenyl)ethylamine 98.0+%, TCI America™

Catalog No. D29255G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D29255G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D29255G Supplier TCI America Supplier No. D29255G

Chemical Identifiers

CAS 52516-13-9
Molecular Formula C8H10Cl2N
Molecular Weight (g/mol) 191.07
MDL Number MFCD00060611
InChI Key VHJKDOLGYMULOP-UHFFFAOYSA-O
Synonym 2,4-dichlorophenethylamine, 2-2,4-dichlorophenyl ethanamine, 2-2,4-dichlorophenyl ethylamine, 2-2,4-dichlorophenyl ethan-1-amine, benzeneethanamine, 2,4-dichloro, acmc-209kzz, 2,4-dichlorophenethyl amine, ksc497e8j, benzeneethanamine,2,4-dichloro
PubChem CID 142938
IUPAC Name 2-(2,4-dichlorophenyl)ethan-1-aminium
SMILES [NH3+]CCC1=CC=C(Cl)C=C1Cl

Specifications

Color Yellow
Quantity 5 g
Formula Weight 190.07
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-(2,4-Dichlorophenyl)ethylamine