missing translation for 'onlineSavingsMsg'
Learn More

2,2,4,4-Tetramethylpentane 99.0+%, TCI America™

Catalog No. p-7136920
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 mL
5 mL
Catalog No. Quantity
T17045ML 5 mL
T170425ML 25 mL
2 options

Catalog No. T17045ML

Supplier: TCI America T17045ML

Only null left

Chemical Identifiers

CAS 1070-87-7
Molecular Formula C9H20
Molecular Weight (g/mol) 128.26
MDL Number MFCD00060873
InChI Key GUMULFRCHLJNDY-UHFFFAOYSA-N
PubChem CID 14058
IUPAC Name 2,2,4,4-tetramethylpentane
SMILES CC(C)(C)CC(C)(C)C

Specifications

Color Colorless
Boiling Point 122°C
Quantity 5 mL
UN Number 1920
Synonym di-tert-butylmethane, pentane, 2,2,4,4-tetramethyl, unii-7js6eal3ee, 7js6eal3ee, pentane, 2,2,4,4-tetramethyl-8ci 9ci, 2,4,4-tetramethylpentane, acmc-2098nd, pentane,2,4,4-tetramethyl, 2,2,4,4,-tetramethylpentane, pentane,2,2,4,4-tetramethyl
Formula Weight 128.26
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,2,4,4-Tetramethylpentane
EINECSNumber (2)-0009
TSCA No