missing translation for 'onlineSavingsMsg'
Learn More

2,2,4,4-Tetramethylpentane, 98%

Catalog No. AAB2164218
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2164218 50 g
AAB2164209 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2164218 Supplier Thermo Scientific Chemicals Supplier No. B2164218
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1070-87-7 | C9H20 | 128.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1070-87-7
Molecular Formula C9H20
Molecular Weight (g/mol) 128.26
MDL Number MFCD00060873
InChI Key GUMULFRCHLJNDY-UHFFFAOYSA-N
PubChem CID 14058
IUPAC Name 2,2,4,4-tetramethylpentane
SMILES CC(C)(C)CC(C)(C)C

Specifications

Density 0.72
Boiling Point 121°C
Flash Point 6°C (43°F)
Odor Petroleum-like
Linear Formula (CH3)3CCH2C(CH3)3
Quantity 50 g
UN Number UN3295
Synonym di-tert-butylmethane, pentane, 2,2,4,4-tetramethyl, unii-7js6eal3ee, 7js6eal3ee, pentane, 2,2,4,4-tetramethyl-8ci 9ci, 2,4,4-tetramethylpentane, acmc-2098nd, pentane,2,4,4-tetramethyl, 2,2,4,4,-tetramethylpentane, pentane,2,2,4,4-tetramethyl
Formula Weight 128.26
Percent Purity 98%
Chemical Name or Material 2,2,4,4-Tetramethylpentane
Show More Show Less
Hazard Category H225-H304
Hazard Statement GHS H Statement
H225
Highly flammable liquid and vapour.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P301+P310-P303+P361+P353-P331-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only