missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,2,4,4-Tetramethyl-1,3-cyclobutanediol (mixture of isomers) 98.0+%, TCI America™
Supplier: TCI America T014525G
Specifications
| 2,2,4,4-Tetramethyl-1,3-cyclobutanediol (mixture of isomers) | |
| 127°C | |
| C8H16O2 | |
| 25 g | |
| FQXGHZNSUOHCLO-UHFFFAOYSA-N | |
| 2,2,4,4-tetramethylcyclobutane-1,3-diol | |
| 76382 | |
| ≥98.0% (GC) |
| 3010-96-6 | |
| White | |
| MFCD00001329 | |
| 2,2,4,4-tetramethyl-1,3-cyclobutanediol, 1,3-cyclobutanediol, 2,2,4,4-tetramethyl, tetramethyl-1,3-cyclobutanediol, 1,1,3,3-tetramethylcyclobutanediol, unii-v7wk40i307, cis-2,2,4,4-tetramethyl-1,3-cyclobutanediol, 2,2,4,4-tetramethylcyclobutane-1,3-diol, mixed isomers, 2,2,4,4-tetramethyl-1,3-cyclobutanediol, mixture of isomers, 2,2,4,4-tetramethylcyclobutane-1,3-diol, cis, acmc-209hd8 | |
| CC1(C(C(C1O)(C)C)O)C | |
| 144.214 | |
| 144.21 | |
| Crystalline Powder |
Chemical Identifiers
| 3010-96-6 | |
| 144.214 | |
| FQXGHZNSUOHCLO-UHFFFAOYSA-N | |
| 76382 | |
| CC1(C(C(C1O)(C)C)O)C |
| C8H16O2 | |
| MFCD00001329 | |
| 2,2,4,4-tetramethyl-1,3-cyclobutanediol, 1,3-cyclobutanediol, 2,2,4,4-tetramethyl, tetramethyl-1,3-cyclobutanediol, 1,1,3,3-tetramethylcyclobutanediol, unii-v7wk40i307, cis-2,2,4,4-tetramethyl-1,3-cyclobutanediol, 2,2,4,4-tetramethylcyclobutane-1,3-diol, mixed isomers, 2,2,4,4-tetramethyl-1,3-cyclobutanediol, mixture of isomers, 2,2,4,4-tetramethylcyclobutane-1,3-diol, cis, acmc-209hd8 | |
| 2,2,4,4-tetramethylcyclobutane-1,3-diol |
Safety and Handling
TSCA : Yes