missing translation for 'onlineSavingsMsg'
Learn More

2,2,4,4-Tetramethyl-1,3-cyclobutanediol (mixture of isomers) 98.0+%, TCI America™

Catalog No. T01455G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T01455G 5 g
T014525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T01455G Supplier TCI America Supplier No. T01455G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 3010-96-6
Molecular Formula C8H16O2
Molecular Weight (g/mol) 144.214
MDL Number MFCD00001329
InChI Key FQXGHZNSUOHCLO-UHFFFAOYSA-N
Synonym 2,2,4,4-tetramethyl-1,3-cyclobutanediol, 1,3-cyclobutanediol, 2,2,4,4-tetramethyl, tetramethyl-1,3-cyclobutanediol, 1,1,3,3-tetramethylcyclobutanediol, unii-v7wk40i307, cis-2,2,4,4-tetramethyl-1,3-cyclobutanediol, 2,2,4,4-tetramethylcyclobutane-1,3-diol, mixed isomers, 2,2,4,4-tetramethyl-1,3-cyclobutanediol, mixture of isomers, 2,2,4,4-tetramethylcyclobutane-1,3-diol, cis, acmc-209hd8
PubChem CID 76382
IUPAC Name 2,2,4,4-tetramethylcyclobutane-1,3-diol
SMILES CC1(C(C(C1O)(C)C)O)C

Specifications

Melting Point 127°C
Color White
Quantity 5 g
Formula Weight 144.21
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,2,4,4-Tetramethyl-1,3-cyclobutanediol (mixture of isomers)