missing translation for 'onlineSavingsMsg'
Learn More

2,2,4,4,6,6-Hexamethylcyclotrisilazane 97.0+%, TCI America™

Catalog No. p-7130801
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H09095ML 5 mL
H090925ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H09095ML Supplier TCI America Supplier No. H09095ML

Chemical Identifiers

CAS 1009-93-4
Molecular Formula C6H21N3Si3
Molecular Weight (g/mol) 219.51
MDL Number MFCD00039698
InChI Key WGGNJZRNHUJNEM-UHFFFAOYSA-N
Synonym Dimethylsilazane Cyclic Trimer
PubChem CID 66094
IUPAC Name 2,2,4,4,6,6-hexamethyl-1,3,5,2,4,6-triazatrisilinane
SMILES C[Si]1(N[Si](N[Si](N1)(C)C)(C)C)C

Specifications

Color Colorless
Boiling Point 188°C
Quantity 5 mL
UN Number 1760
Formula Weight 219.51
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2,2,4,4,6,6-Hexamethylcyclotrisilazane
EINECSNumber (5)-6044
RTECSNumber GZ4720000
TSCA Yes