missing translation for 'onlineSavingsMsg'
Learn More

2,2,4,4,6,6-Hexamethyl-1,3,5-trithiane 98.0+%, TCI America™

Catalog No. h12785g
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H12785G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H12785G Supplier TCI America Supplier No. H12785G
Only null left

Chemical Identifiers

CAS 828-26-2
Molecular Formula C9H18S3
Molecular Weight (g/mol) 222.42
MDL Number MFCD00066341
InChI Key NBNWHQAWKFYFKI-UHFFFAOYSA-N
Synonym trithioacetone, 2,2,4,4,6,6-hexamethyl-s-trithiane, hexamethyl-1,3,5-trithiane, hexamethyl-s-trithiane, 1,3,5-trithiane, 2,2,4,4,6,6-hexamethyl, s-trithiane, hexamethyl, unii-l0me5b2zwy, fema no. 3475, s-trithiane, 2,2,4,4,6,6-hexamethyl, l0me5b2zwy
PubChem CID 13233
IUPAC Name hexamethyl-1,3,5-trithiane
SMILES CC1(C)SC(C)(C)SC(C)(C)S1

Specifications

Color Yellow
Boiling Point 107°C
Quantity 5 g
Formula Weight 222.42
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,2,4,4,6,6-Hexamethyl-1,3,5-trithiane
RTECSNumber YL8350000
TSCA Yes