missing translation for 'onlineSavingsMsg'
Learn More

2,2,3-Trimethylbutane 96.0+%, TCI America™

Catalog No. T07115ML
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T07115ML 5 mL
T071125ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T07115ML Supplier TCI America Supplier No. T07115ML

Chemical Identifiers

CAS 464-06-2
Molecular Formula C7H16
Molecular Weight (g/mol) 100.205
MDL Number MFCD00039846
InChI Key ZISSAWUMDACLOM-UHFFFAOYSA-N
Synonym triptan, triptane, butane, 2,2,3-trimethyl, unii-40v943jdgr, 2,3-trimethylbutane, butane,2,3-trimethyl, 2,2,3-trimethyl-butane, acmc-1ahi7, butane,2,2,3-trimethyl, 2,3,3-trimethylbutan-2-yl
PubChem CID 10044
IUPAC Name 2,2,3-trimethylbutane
SMILES CC(C)C(C)(C)C

Specifications

Melting Point -24°C
Color Colorless
Boiling Point 81°C
Quantity 5 mL
UN Number 1206
Formula Weight 100.21
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,2,3-Trimethylbutane
EINECSNumber (2)-0007
TSCA Yes