Learn More
2,2,3-Trimethylbutane, 99%, Thermo Scientific Chemicals
$169.20 - $169.20
Chemical Identifiers
| CAS | 464-06-2 |
|---|---|
| Molecular Formula | C7H16 |
| Molecular Weight (g/mol) | 100.2 |
| MDL Number | MFCD00039846 |
| InChI Key | ZISSAWUMDACLOM-UHFFFAOYSA-N |
| Synonym | triptan, triptane, butane, 2,2,3-trimethyl, unii-40v943jdgr, 2,3-trimethylbutane, butane,2,3-trimethyl, 2,2,3-trimethyl-butane, acmc-1ahi7, butane,2,2,3-trimethyl, 2,3,3-trimethylbutan-2-yl |
| PubChem CID | 10044 |
| IUPAC Name | 2,2,3-trimethylbutane |
| SMILES | CC(C)C(C)(C)C |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC398620050
|
Thermo Scientific Chemicals
398620050 |
5 g | Glass bottle |
Each for $169.20
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 464-06-2 | |
| 100.2 | |
| ZISSAWUMDACLOM-UHFFFAOYSA-N | |
| 10044 | |
| CC(C)C(C)(C)C |
| C7H16 | |
| MFCD00039846 | |
| triptan, triptane, butane, 2,2,3-trimethyl, unii-40v943jdgr, 2,3-trimethylbutane, butane,2,3-trimethyl, 2,2,3-trimethyl-butane, acmc-1ahi7, butane,2,2,3-trimethyl, 2,3,3-trimethylbutan-2-yl | |
| 2,2,3-trimethylbutane |
Specifications
| 464-06-2 | |
| 100.0 | |
| Colorless | |
| 81.0°C | |
| Authentic | |
| Glass bottle | |
| 1.3880 to 1.3900 (20°C, 589nm) | |
| MFCD00039846 | |
| 0.69 | |
| ZISSAWUMDACLOM-UHFFFAOYSA-N | |
| 2,2,3-trimethylbutane | |
| 10044 | |
| 99% | |
| Liquid |
| 98.5 | |
| -25.0°C | |
| 0.6900g/mL | |
| −7°C | |
| 98.5% min. (GC) | |
| C7H16 | |
| CH3C(CH3)2CH(CH3)2 | |
| 5 g | |
| triptan, triptane, butane, 2,2,3-trimethyl, unii-40v943jdgr, 2,3-trimethylbutane, butane,2,3-trimethyl, 2,2,3-trimethyl-butane, acmc-1ahi7, butane,2,2,3-trimethyl, 2,3,3-trimethylbutan-2-yl | |
| CC(C)C(C)(C)C | |
| 100.2 | |
| 100.2 | |
| Extra Pure | |
| 2, 2, 3-Trimethylbutane |
Safety and Handling
GHS H Statement:
May cause drowsiness or dizziness.
Very toxic to aquatic life with long lasting effects.
May be fatal if swallowed and enters airways.
Causes skin irritation.
Highly flammable liquid and vapor.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
EINECSNumber : 207-346-3
RUO – Research Use Only