missing translation for 'onlineSavingsMsg'
Learn More

2,2,3-Trimethylbutane, 99%, Thermo Scientific Chemicals

Catalog No. AC398620050
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC398620050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC398620050 Supplier Thermo Scientific Chemicals Supplier No. 398620050

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 464-06-2
Molecular Formula C7H16
Molecular Weight (g/mol) 100.2
MDL Number MFCD00039846
InChI Key ZISSAWUMDACLOM-UHFFFAOYSA-N
Synonym triptan, triptane, butane, 2,2,3-trimethyl, unii-40v943jdgr, 2,3-trimethylbutane, butane,2,3-trimethyl, 2,2,3-trimethyl-butane, acmc-1ahi7, butane,2,2,3-trimethyl, 2,3,3-trimethylbutan-2-yl
PubChem CID 10044
IUPAC Name 2,2,3-trimethylbutane
SMILES CC(C)C(C)(C)C

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -25.0°C
Color Colorless
Density 0.6900g/mL
Boiling Point 81.0°C
Flash Point −7°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.3880 to 1.3900 (20°C, 589nm)
Linear Formula CH3C(CH3)2CH(CH3)2
Quantity 5 g
Specific Gravity 0.69
Formula Weight 100.2
Percent Purity 99%
Grade Extra Pure
Physical Form Liquid
Chemical Name or Material 2, 2, 3-Trimethylbutane
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
May cause drowsiness or dizziness.
Very toxic to aquatic life with long lasting effects.
May be fatal if swallowed and enters airways.
Causes skin irritation.
Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 207-346-3

RUO – Research Use Only