missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ 2,2,3-Trimethylbutane, 98%

Catalog No. AAB2020503
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2020503 1 g
AAB2020506 5 g
AAB2020514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2020503 Supplier Thermo Scientific™ Supplier No. B2020503
Only null left

CAS: 464-06-2 | C7H16 | 100.21 g/mol

Chemical Identifiers

CAS 464-06-2
Molecular Weight (g/mol) 100.21
MDL Number MFCD00039846
InChI Key ZISSAWUMDACLOM-UHFFFAOYSA-N
IUPAC Name 2,2,3-trimethylbutane
SMILES CC(C)C(C)(C)C

Specifications

Melting Point -25°
Density 0.690
Boiling Point 80-82°
Flash Point -6°(20°F)
Molecular Formula C7H16
Linear Formula (CH{3}){3}CCH(CH{3}){2}
Quantity 1 g
UN Number UN1206
Formula Weight 100.21
Physical Form Liquid
ShelfLife 5 Years
EINECSNumber 207-346-3