missing translation for 'onlineSavingsMsg'
Learn More

2,2,3,4,4,4-Hexafluorobutyl Methacrylate (stabilized with MEHQ) 98.0+%, TCI America™

Catalog No. p-7131109
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H15555G 5 g
H155525G 25 g
H1555100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. H15555G Supplier TCI America Supplier No. H15555G

Chemical Identifiers

CAS 36405-47-7
Molecular Formula C8H8F6O2
Molecular Weight (g/mol) 250.14
MDL Number MFCD00042311
InChI Key DFVPUWGVOPDJTC-UHFFFAOYSA-N
Synonym 2,2,3,4,4,4-hexafluorobutyl methacrylate, 1h,1h,3h-hexafluorobutyl methacrylate, hexafluorobutyl methacrylate, methacrylic acid 2,2,3,4,4,4-hexafluorobutyl ester, 2-propenoic acid, 2-methyl-, 2,2,3,4,4,4-hexafluorobutyl ester, hexafluorobutyl methacrylate polymer, hfbma, acmc-1adv6, ksc489s1j, 2-propenoic acid, 2-methyl-, 2,2,3,4,4,4-hexafluorobutyl ester, homopolymer
PubChem CID 549772
IUPAC Name 2,2,3,4,4,4-hexafluorobutyl 2-methylprop-2-enoate
SMILES CC(=C)C(=O)OCC(C(C(F)(F)F)F)(F)F

Specifications

Color Colorless
Boiling Point 158°C
Quantity 5 g
UN Number 3272
Formula Weight 250.14
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,2,3,4,4,4-Hexafluorobutyl Methacrylate (stabilized with MEHQ)