missing translation for 'onlineSavingsMsg'
Learn More

2,2,3,4,4,4-Hexafluorobutyl methacrylate, 96%, stab.

Catalog No. AAL0954314
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0954314 25 g
AAL0954322 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0954314 Supplier Thermo Scientific Chemicals Supplier No. L0954314
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 36405-47-7 | C8H8F6O2 | 250.14 g/mol

It is an epoxy thinner and is mainly used as the modifier of the (meth)acrylate resin. Well-defined organic/inorganic hybrid fluorinated star polymers were synthesized via atom transfer radical polymerization (ATRP) of 2,2,3,4,4,4-hexafluorobutyl methacrylate (HFBMA) using octa(aminophenyl)silsesquioxane (OAPS) nano-cage as initiator.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an epoxy thinner and is mainly used as the modifier of the (meth)acrylate resin. Well-defined organic/inorganic hybrid fluorinated star polymers were synthesized via atom transfer radical polymerization (ATRP) of 2,2,3,4,4,4-hexafluorobutyl methacrylate (HFBMA) using octa(aminophenyl)silsesquioxane (OAPS) nano-cage as initiator.

Solubility
Difficult to mix in water.

Notes
Store at room temperature. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 36405-47-7
Molecular Formula C8H8F6O2
Molecular Weight (g/mol) 250.14
MDL Number MFCD00042311
InChI Key DFVPUWGVOPDJTC-UHFFFAOYSA-N
Synonym 2,2,3,4,4,4-hexafluorobutyl methacrylate, 1h,1h,3h-hexafluorobutyl methacrylate, hexafluorobutyl methacrylate, methacrylic acid 2,2,3,4,4,4-hexafluorobutyl ester, 2-propenoic acid, 2-methyl-, 2,2,3,4,4,4-hexafluorobutyl ester, hexafluorobutyl methacrylate polymer, hfbma, acmc-1adv6, ksc489s1j, 2-propenoic acid, 2-methyl-, 2,2,3,4,4,4-hexafluorobutyl ester, homopolymer
PubChem CID 549772
IUPAC Name 2,2,3,4,4,4-hexafluorobutyl 2-methylprop-2-enoate
SMILES CC(=C)C(=O)OCC(C(C(F)(F)F)F)(F)F

Specifications

Density 1.352
Boiling Point 158°C
Flash Point 56°C (132°F)
Refractive Index 1.362
Quantity 25 g
UN Number UN3272
Beilstein 2725177
Solubility Information Difficult to mix in water.
Formula Weight 250.14
Percent Purity 96%
Chemical Name or Material 2,2,3,4,4,4-Hexafluorobutyl methacrylate, stabilized
Show More Show Less
Hazard Category H226
Hazard Statement GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ESTERS, N.O.S.
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only