missing translation for 'onlineSavingsMsg'
Learn More

2,2,3,3-Tetrafluoro-1-propanol 98.0+%, TCI America™

Catalog No. T010125G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T010125G 25 g
T0101100G 100 g
T0101500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T010125G Supplier TCI America Supplier No. T010125G

Chemical Identifiers

CAS 76-37-9
Molecular Formula C3H4F4O
Molecular Weight (g/mol) 132.06
MDL Number MFCD00004676
InChI Key NBUKAOOFKZFCGD-UHFFFAOYSA-N
Synonym 2,2,3,3-tetrafluoro-1-propanol, 2,2,3,3-tetrafluoropropanol, 1h,1h,3h-tetrafluoro-1-propanol, 1-propanol, 2,2,3,3-tetrafluoro, tetrafluoropropyl alcohol, 2,2,3,3-tetrafluoropropyl alcohol, 1,1,3-trihydroperfluoro-1-propanol, unii-rno513716p, 1h,1h,3h-tetrafluoropropanol, wln: q1xffyff
PubChem CID 6441
IUPAC Name 2,2,3,3-tetrafluoropropan-1-ol
SMILES OCC(F)(F)C(F)F

Specifications

Color Colorless
Boiling Point 106°C
Quantity 25 g
UN Number 1993
Formula Weight 132.06
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,2,3,3-Tetrafluoro-1-propanol
EINECSNumber (2)-0284
RTECSNumber UB9400000
TSCA Yes