missing translation for 'onlineSavingsMsg'
Learn More

2,2,3,3-Tetrafluoro-1,4-butanediol, 97%

Catalog No. p-7046869
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1663503 1 g
AAL1663506 5 g
AAL1663514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL1663503 Supplier Thermo Scientific Chemicals Supplier No. L1663503

CAS: 425-61-6 | C4H6F4O2 | 162.08 g/mol

2,2,3,3-Tetrafluoro-1,4-butanediol is a reactant used for the enzymic synthesis of silicone fluorinated aliphatic polyester amides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2,3,3-Tetrafluoro-1,4-butanediol is a reactant used for the enzymic synthesis of silicone fluorinated aliphatic polyester amides.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with acids and oxidizing agents.

Chemical Identifiers

CAS 425-61-6
Molecular Formula C4H6F4O2
Molecular Weight (g/mol) 162.08
MDL Number MFCD00042375
InChI Key CDZXJJOGDCLNKX-UHFFFAOYSA-N
Synonym 2,2,3,3-tetrafluoro-1,4-butanediol, 1,4-dihydroxy-2,2,3,3-tetrafluorobutane, acmc-1aqx7, 1,4-butanediol,2,2,3,3-tetrafluoro, 2,2,3,3-tetrakis fluoranyl butane-1,4-diol
PubChem CID 136270
IUPAC Name 2,2,3,3-tetrafluorobutane-1,4-diol
SMILES OCC(F)(F)C(F)(F)CO

Specifications

Melting Point 80°C to 87°C
Boiling Point 110°C to 112°C (13 mmHg)
Quantity 1 g
Beilstein 1745847
Solubility Information Soluble in water.
Formula Weight 162.08
Percent Purity 97%
Chemical Name or Material 2,2,3,3-Tetrafluoro-1,4-butanediol
Hazard Category H315-H319
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only