missing translation for 'onlineSavingsMsg'
Learn More

2,2,3,3,4,4,5,5-Octafluoro-1,6-hexanediol 98.0+%, TCI America™

Catalog No. p-7134282
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O02945G 5 g
O029425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O02945G Supplier TCI America Supplier No. O02945G
Only null left

Chemical Identifiers

CAS 355-74-8
Molecular Formula C6H6F8O2
Molecular Weight (g/mol) 262.099
MDL Number MFCD00069087
InChI Key NHEKBXPLFJSSBZ-UHFFFAOYSA-N
Synonym 2,2,3,3,4,4,5,5-octafluoro-1,6-hexanediol, 1,6-dihydroxy-2,2,3,3,4,4,5,5-octafluorohexane, 1,6-hexanediol, 2,2,3,3,4,4,5,5-octafluoro, 2,2,3,3,4,4,5,5-octafluorohexan-1,6-diol, acmc-1code, nhekbxplfjssbz-uhfffaoysa, 2,2,3,3,4,4,5,5octafluoro-1,6-hexanediol
PubChem CID 136181
IUPAC Name 2,2,3,3,4,4,5,5-octafluorohexane-1,6-diol
SMILES C(C(C(C(C(CO)(F)F)(F)F)(F)F)(F)F)O

Specifications

Melting Point 68°C
Quantity 5 g
Formula Weight 262.10
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,2,3,3,4,4,5,5-Octafluoro-1,6-hexanediol