missing translation for 'onlineSavingsMsg'
Learn More

2,2,3,3,4,4,5,5-Octafluoro-1,6-hexanediol, 97%

Catalog No. p-7046844
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1656006 5 g
AAL1656014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1656006 Supplier Thermo Scientific Chemicals Supplier No. L1656006
Only null left

CAS: 355-74-8 | C6H6F8O2 | 262.099 g/mol

2,2,3,3,4,4,5,5-Octafluoro-1,6-hexanediol is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2,3,3,4,4,5,5-Octafluoro-1,6-hexanediol is used as a pharmaceutical intermediate.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 355-74-8
Molecular Formula C6H6F8O2
Molecular Weight (g/mol) 262.099
MDL Number MFCD00069087
InChI Key NHEKBXPLFJSSBZ-UHFFFAOYSA-N
Synonym 2,2,3,3,4,4,5,5-octafluoro-1,6-hexanediol, 1,6-dihydroxy-2,2,3,3,4,4,5,5-octafluorohexane, 1,6-hexanediol, 2,2,3,3,4,4,5,5-octafluoro, 2,2,3,3,4,4,5,5-octafluorohexan-1,6-diol, acmc-1code, nhekbxplfjssbz-uhfffaoysa, 2,2,3,3,4,4,5,5octafluoro-1,6-hexanediol
PubChem CID 136181
IUPAC Name 2,2,3,3,4,4,5,5-octafluorohexane-1,6-diol
SMILES C(C(C(C(C(CO)(F)F)(F)F)(F)F)(F)F)O

Specifications

Melting Point 67°C to 70°C
Boiling Point 100°C (3 mmHg)
Quantity 5 g
Beilstein 1784039
Solubility Information Soluble in water.
Formula Weight 262.1
Percent Purity 97%
Chemical Name or Material 2,2,3,3,4,4,5,5-Octafluoro-1,6-hexanediol
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only