missing translation for 'onlineSavingsMsg'
Learn More

2,2,3,3,4,4,5,5,6,6,7,7-Dodecafluoroheptyl Acrylate (stabilized with TBC) 97.0+%, TCI America™

Catalog No. D49895G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D49895G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D49895G Supplier TCI America Supplier No. D49895G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2993-85-3
Molecular Formula C10H6F12O2
Molecular Weight (g/mol) 386.137
MDL Number MFCD00080746
InChI Key QJEJDNMGOWJONG-UHFFFAOYSA-N
Synonym 1h,1h,7h-dodecafluoroheptyl acrylate, 2,2,3,3,4,4,5,5,6,6,7,7-dodecafluoroheptyl acrylate, 2-propenoic acid, 2,2,3,3,4,4,5,5,6,6,7,7-dodecafluoroheptyl ester, 2-propenoic acid,2,2,3,3,4,4,5,5,6,6,7,7-dodecafluoroheptyl ester, dfha, 1h,1h,7h perfluoroheptyl acrylate, 1h,1h,7h-perfluoroheptyl acrylate, acrylic acid 2,2,3,3,4,4,5,5,6,6,7,7-dodecafluoroheptyl ester, 2,2,3,3,4,4,5,5,6,6,7,7-dodecafluoroheptyl acrylate stabilized with tbc
PubChem CID 76340
IUPAC Name 2,2,3,3,4,4,5,5,6,6,7,7-dodecafluoroheptyl prop-2-enoate
SMILES C=CC(=O)OCC(C(C(C(C(C(F)F)(F)F)(F)F)(F)F)(F)F)(F)F

Specifications

Color Colorless
Boiling Point 107°C
Quantity 5 g
Formula Weight 386.14
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,2,3,3,4,4,5,5,6,6,7,7-Dodecafluoroheptyl Acrylate (stabilized with TBC)
RTECSNumber UD3340500
TSCA No
Recommended Storage Refrigerator